EMAIL THIS PAGE TO A FRIEND
Journal of medical entomology
The growth and serial passage of cell lines from Aedes aegypti (L.) larvae in different media.
PMID 5360492
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90120514
TRA-171