EMAIL THIS PAGE TO A FRIEND
Journal of Analytical Atomic Spectrometry
Determination of cadmium by flow injection atomic absorption spectrometry with cold vapor generation by a tetrahydroborate-form anion-exchanger.
PMID 542571