EMAIL THIS PAGE TO A FRIEND
Experimental cell research
Mouse myelomas and lymphomas in culture.
PMID 5439579
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85011401
P3X63Ag8