EMAIL THIS PAGE TO A FRIEND
The Journal of laboratory and clinical medicine
Studies on a procoagulant fraction of southern copperhead snake venom: the preferential release of fibrinopeptide B.
PMID 5458334