EMAIL THIS PAGE TO A FRIEND
FEBS letters
Growth-modulating human plasma tripeptide: relationship between molecular structure and DNA synthesis in hepatoma cells.
PMID 556547
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1887
Gly-His-Lys acetate salt, ≥97% (TLC)
C16H28N6O6