EMAIL THIS PAGE TO A FRIEND
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. 1. Abt. Medizinisch-hygienische Bakteriologie, Virusforschung und Parasitologie. Originale
[Selective culture media with egg yolk for the isolation of pathogenic staphylococci from foods].
PMID 5633613
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60985 KRANEP Agar, for microbiology