EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
An automated fluorometric assay for alkaline phosphatase using 3-O-methylfluorescein phosphate.
PMID 5665203
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2629
3-O-Methylfluorescein phosphate cyclohexylammonium salt
C21H15O8P · C6H13N