EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Transplantable Leydig cell tumors in Fischer rats: horomone responsivity and hormone production.
PMID 5688092
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89031604
LC540