EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Acetylcholinesterase hydrolysis of halogen substituted acetylcholines.
PMID 5689803
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B121
Bromoacetylcholine bromide
C7H15BrNO2 · Br