EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Comparative studies on the inhibition of trypsin, plasmin, and thrombin by derivatives of benzylamine and benzamidine.
PMID 5701967
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06880
4-Aminobenzamidine dihydrochloride, BioReagent, suitable for fluorescence, ≥99.0% (TLC)
C7H9N3 · 2HCl