EMAIL THIS PAGE TO A FRIEND
Biochemistry
The oxygenation of hemoglobin in the presence of 2,3-diphosphoglycerate. Effect of temperature, pH, ionic strength, and hemoglobin concentration.
PMID 5799137
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5764
2,3-Diphospho-D-glyceric acid pentasodium salt, glycolysis metabolite
C3H3Na5O10P2