EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
The synthesis of beta-carboline derivatives. VII. The isolation of the possible intermediate in the condensation of 3-(2-bromoethyl)indole and 2-halogenopyridine.
PMID 5867834
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
376523
3-(2-Bromoethyl)indole, 97%
C10H10BrN