EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis of heterocyclic-substituted chromones and related compounds as potential anticancer agents.
PMID 5885091
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
525928
Quinaldoyl chloride, 97%
C10H6ClNO