EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
A new mast cell degranulating peptide, granuliberin-R, in the frog (Rana rugosa) skin.
PMID 589733
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7897
Granuliberin R, ≥97% (HPLC)
C69H103N19O14