EMAIL THIS PAGE TO A FRIEND
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.)
Isolation, propagation and neutralization of rubella virus in cultures of rabbit cornea (SIRC) cells.
PMID 5918951
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89090404
SIRC