EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Inactivation of rabbit muscle adenosine triphosphate-adenosine 5'-phosphate phosphotransferase by alkylation of methionine residues.
PMID 6017726