EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Dipeptidyl arylamidase III of the pituitary. Purification and characterization.
PMID 6061409
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3261
Arg-Arg β-naphthylamide trihydrochloride
C22H33N9O2 · 3HCl