EMAIL THIS PAGE TO A FRIEND
Helvetica chimica acta
[The metabolic products of microorganisms. Boromycin].
PMID 6081908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59770
(−)-Isopulegol, analytical standard
C10H18O