EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Opiate activity and receptor selectivity of dynorphin1-13 and related peptides.
PMID 6133254
Abstract
Experiments on the mouse vas deferens preparation obtained from animals chronically pretreated with selective opioid agonists have revealed the existence of opiate receptors with a high degree of preference for dynorphin1-13, alpha-neoendorphin1-8 and the eta-agonist MR 2034. Modification of the dynorphin sequence, by introduction of D-alanine into position 2, resulted in a complete loss of dynorphin-like agonistic activity on the tolerant mouse vas deferens. None of the dynorphin-related peptides tested displayed a pronounced opiate-like antinociception when administered intracerebroventricularly into rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9148
[D-Arg6]-Dynorphin A Porcine fragment 1-13, ≥97% (HPLC)
C75H126N24O15