EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The kinetic mechanism of ox liver glutamate dehydrogenase in the presence of the allosteric effector ADP. The oxidative deamination of L-glutamate.
PMID 6149744
Abstract
In steady-state kinetic studies of ox liver glutamate dehydrogenase in 0.11 M-potassium phosphate buffer, pH7, at 25 degrees C, the concentration of ADP was varied from 0.5 to 1000 microM. Inhibition was observed except when the concentrations of both glutamate and coenzyme were high, when activation was seen. With NAD+ or NADP+ as coenzyme, 200 microM-ADP was sufficient to saturate the enzyme with respect to the major effect of this nucleotide. In the presence of 210 microM-ADP, widely varied concentrations of coenzyme give linear Lineweaver-Burk plots, in marked contrast with results obtained previously for kinetics without ADP. This has allowed evaluation for the reaction with NAD+, NADP+ and acetylpyridine-adenine dinucleotide (315 microM-ADP in the last case) of all four initial rate parameters, i.e. the phi coefficients in the equation: (Formula: see text) where A is coenzyme and B is glutamate. The relative constancy of phi B and of phi AB/phi A with the different coenzymes point to a compulsory-order mechanism with glutamate as the leading substrate. This conclusion, though unexpected, agrees well with various previous observations on the binding of oxidized coenzyme.