EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
2-[[[2-(2,6-Dimethoxyphenoxy)ethyl]amino]-methyl] -1,4-benzoxathian: a new antagonist with high potency and selectivity toward alpha 1-adrenoreceptors.
PMID 6150111
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B016
Benoxathian hydrochloride, solid
C19H23NO4S · HCl