EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Fractionation of low molecular weight DNA or RNA in polyacrylamide gels containing 98% formamide or 7 M urea.
PMID 6154865
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51456
Urea, BioUltra, for molecular biology, ≥99.5% (T)
CH4N2O