EMAIL THIS PAGE TO A FRIEND
Biochemistry
Effect of ribonucleoside-vanadyl complexes on enzyme-catalyzed reactions central to recombinant deoxyribonucleic acid technology.
PMID 6182903
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3380 Ribonucleoside vanadyl complexes, 200 mM