EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Solid phase radioimmunoassay for human myelin basic protein using a monoclonal antibody.
PMID 6194177
Abstract
A solid phase competitive assay for human myelin basic protein (MBP) has been developed using a monoclonal antibody to MBP. The assay has been applied to the detection of antigenic peptides derived from MBP, the measurement of anti-idiotypic antibody and the detection of MBP in human serum.