EMAIL THIS PAGE TO A FRIEND
Endocrinology
Growth and function of cultured bovine adrenocortical cells in a serum-free defined medium.
PMID 6213403
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91021201
SBAC