EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Opioid hexapeptides and heptapeptides in adrenal medulla and brain possible implications on the biosynthesis of enkephalins.
PMID 6258484