EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and antiviral properties of (Z)-5-(2-bromovinyl)-2'-deoxyuridine.
PMID 6265638
Abstract
(Z)-5-(2-Bromovinyl)uracil was obtained by photoisomerization of the E. isomer. Similarly, (E)-5-(2-bromovinyl)-2'-deoxyuridine gave the required Z isomer. (Z)-5-(2-Bromovinyl)-2'-deoxyuridine is much less active against herpes simplex virus type 1 (HSV-1) and somewhat less active against herpes simplex virus type 2 than is the E isomer. Both isomers show similar activity against vaccinia virus. Therefore, the highly potent and selective activity of (E)-5-(2-bromovinyl)-2'-deoxyuridine against HSV-1 is due to its E configuration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457442
(E)-5-(2-Bromovinyl)uracil, 97%
C6H5BrN2O2