EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
[3H]rauwolscine (alpha-yohimbine): a specific antagonist radioligand for brain alpha 2-adrenergic receptors.
PMID 6276200
Abstract
[3H]Rauwolscine, a specific and potent alpha 2-antagonist radioligand, was used to characterize alpha 2-receptor binding in bovine cerebral cortex. [3H]Rauwolscine binding was reversible, stereospecific, and saturable. Association, dissociation, and saturation studies revealed one site interactions (k -1/k+1 = 1.2 nM, KD = 2.5 nM, Bmax = 160 fmol/mg protein) and competition studies indicated that [3H]rauwolscine labeled the alpha 2-receptor. Agonists inhibited [3H]rauwolscine binding in a shallow, GTP-sensitive manner. These results suggest that [3H]rauwolscine specifically labels both the high and low affinity states of the alpha 2-receptor in brain membranes.