EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Dimerization of deoxyribonuclease I, lysozyme and papain. Effects of ionic strength on enzymic activity.
PMID 6282587
Abstract
Transition of bovine ribonuclease A from its monomeric to a dimeric form changes the pattern of enzymic activity response to ionic strength [Sorrentino, S., Carsana, A., Furia, A., Doskocil, J., and Libonati, M. (1980) Biochim. Biophys. Acta. 609, 40-52]. To see whether this phenomenon could be common to other enzyme-substrate systems, the action of various dimeric and monomeric enzymes (ox pancreas deoxyribonuclease, hog spleen acid deoxyribonuclease, bovine seminal ribonuclease, egg-white lysozyme, and papain) on polyelectrolytic substrates has been studied under different conditions of ionic strength. Dimerization of ox pancreas deoxyribonuclease, lysozyme and papain was obtained by cross-linkage with dimethyl suberimidate. The main results of the investigation, similar to those obtained with ribonuclease A, are the following. 1. Enzyme monomers and dimers show markedly different patterns of activity response to ionic strength at given pH values: the reactions catalyzed by monomeric enzymes are highly modulated by salt, whereas those catalyzed by dimeric enzymes are not. In particular, at the reaction optimum the monomeric form of an enzyme is significantly more active than the dimeric one. 2. The optimum of the reaction catalyzed by a dimeric enzyme is shifted to higher ionic strengths in comparison with that of the reaction catalyzed by a monomeric enzyme. A model is proposed that could explain these results on the basis of the influence of ionic strength on the intramolecular dynamics of the enzyme molecule and its non-specific interactions with polyelectrolytic substrates.