EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Glycerol oxidase from Aspergillus japonicus.
PMID 6292659
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
49781
Glycerol solution, 86-89% (T)
C3H8O3 · xH2O