EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Case records of the Massachusetts General Hospital. Weekly clinicopathological exercises. Case 1-1983. A 37-year-old schizophrenic woman with hypoglycemic episode.
PMID 6292720