EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Preparation and characterization of a pentaammineruthenium(III) derivative of horse heart ferricytochrome c.
PMID 6294670
Abstract
A stable complex between pentaammineruthenium(III) and histidine-33 in horse heart ferricytochrome c is formed in the reaction between aquopentaammineruthenium(II) and the protein at pH 7. HPLC of the tryptic hydrolysate of the modified protein was employed to identify the pentaammineruthenium binding site. Spectroscopic measurements show that the integrity of the native structure in the vicinity of the heme c group is maintained in the ruthenium-modified protein. The reduction potentials are: heme c (Fe3+/2+), 0.26 V; Ru(NH3)5(His-33)3+/2+, 0.15 V (vs. normal hydrogen electrode).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G25150 Sephadex® G-25, Medium