EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Agonist and antagonist affinities for inhibitory adenosine receptors are reciprocally affected by 5'-guanylylimidodiphosphate or N-ethylmaleimide.
PMID 6296141