EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Automated kinetic determination of angiotensin-converting enzyme in serum.
PMID 6327119
Abstract
In this automated kinetic modification of a previous method (Anal Biochem 95: 540-548, 1979) for determining angiotensin-converting enzyme (EC 3.4.15.1), 3-(2- furylacryloyl )-L- phenylalanylglycylglycine is used as the substrate. The change in absorbance at 340 nm is used to monitor hydrolysis of the substrate. The rate of hydrolysis is roughly threefold greater than with previously reported substrates, so assay time and sensitivity are improved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7131
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly
C20H21N3O6