EMAIL THIS PAGE TO A FRIEND
Sheng li ke xue jin zhan [Progress in physiology]
[Strychnine discharge and its mechanism].
PMID 6344207
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45661
Strychnine, PESTANAL®, analytical standard
C21H22N2O2