EMAIL THIS PAGE TO A FRIEND
Regulatory toxicology and pharmacology : RTP
Tin--a toxic heavy metal? A review of the literature.
PMID 6371930
Abstract
A tolerable limit for tin concentration in canned food of 250 ppm (Fritsch et al., 1977) is generally accepted. However, biochemical effects attributable to tin have been observed even after oral administration of 1 and 3 mg Sn/kg body wt (Yamaguchi et al., 1980). These doses reflect 10 and 30 ppm tin in the diet. The experiments of de Groot (1973) showed that hemoglobin concentrations in the blood of rats decreased significantly feeding a diet containing 150 ppm tin. The absorption of iron was diminished after simultaneous administration of 0.8 mumol Sn(II) and iron, reflecting a tin dose of 95 ppm tin, by injection into jejunal loops of rats (Schäfer and Forth, 1983). In general, however, canned food usually plays a secondary role in daily nutrition. Fortunately, concentrations of about 2000 ppm tin as reported by Warburton et al. (1962) and Barker and Runte (1972) are not found in canned food, but values between 50 and 500 ppm are not unusual (Piscator, 1979). If a large amount of canned food is eaten daily over a long period, disturbances of gastric acid secretion and a reduction in iron absorption or heme metabolism cannot be excluded. The storage of food, especially acid foods, in opened cans should be avoided as this practice increases the amount of tin in the food when it is consumed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14509
Tin, ≥99%, powder
Sn
265756
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sn
356948
Tin, foil, thickness 0.127 mm, 99.9%
Sn
357316
Tin, foil, thickness 0.1 mm, 99.999%
Sn
265721
Tin, foil, thickness 0.025 mm, 99.999%
Sn
243434
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sn
576883
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sn
265640
Tin, powder, <150 μm, 99.5% trace metals basis
Sn
265632
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sn
520373
Tin, powder, 10 μm, 99% trace metals basis
Sn
204692
Tin, shot, 99.999% trace metals basis
Sn
217697
Tin, 99.8%, shot, 3 mm
Sn
356956
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sn
265691
Tin, wire, diam. 0.25 mm, 99.99%
Sn
755702
Tin, powder, -100 mesh, 99.99% trace metals basis
Sn
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF66339254
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF54760259
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF45567434
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF28221274
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF16505636
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF50720925
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF00846413
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF99151881
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF83193925
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Sn