EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Ion-selective electrodes and their clinical application in the continuous ion monitoring.
PMID 6378025
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30585
Bis(1-butylpentyl)decane-1,10-diyl diglutarate, Selectophore
C38H70O8