EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans
1,8-Dinitropyrene.
PMID 6381287
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284319
1,8-Dinitropyrene, 98%
C16H8N2O4