EMAIL THIS PAGE TO A FRIEND
Clinics in rheumatic diseases
Isoxicam.
PMID 6391795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I1762
Isoxicam, analytical standard
C14H13N3O5S