EMAIL THIS PAGE TO A FRIEND
Seminars in thrombosis and hemostasis
Methodologic and basic aspects of indium-111 platelets.
PMID 6408738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57083
Indium, bars, 99.95%
In
264113
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
In
326607
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
In
357278
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
In
326631
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
In
357286
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
In
264040
Indium, foil, thickness 0.25 mm, 99.998% trace metals basis
In
357294
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
In
264059
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
In
357308
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
In
326615
Indium, pieces, 99.99% trace metals basis
In
203432
Indium, powder, −60 mesh, 99.999% trace metals basis
In
264032
Indium, powder, −100 mesh, 99.99% trace metals basis
In
277959
Indium, powder, 99.99% trace metals basis
In
385379
Indium, rod, diam. 10 mm, 99.9998% trace metals basis
In
264091
Indium, rod, diam. 6.0 mm, ≥99.999% trace metals basis
In
357065
Indium, wire, diam. 2.0 mm, 99.999% trace metals basis
In
278319
Indium, wire, diam. 2.0 mm, 99.99% trace metals basis
In
340863
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
In
357073
Indium, wire, diam. 1.0 mm, 99.999% trace metals basis
In
264075
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
In
264067
Indium, wire, diam. 0.5 mm, 99.999% trace metals basis
In
326623
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
In
GF57106403
Indium, foil, 4mm disks, thickness 0.015mm, 99.8%
In
GF38968666
Indium, foil, 4mm disks, thickness 0.01mm, 99.8%
In
GF29602571
Indium, foil, 4mm disks, thickness 0.025mm, 99.8%
In
GF49944088
Indium, foil, 4mm disks, thickness 0.03mm, 99.8%
In
GF38467301
Indium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.999%
In
GF04865564
Indium, foil, 100x100mm, thickness 0.1mm, as rolled, 100%
In
GF40076276
Indium, foil, 100x100mm, thickness 0.20mm, as rolled, 99.999%
In