EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Purification and sequence analysis of bioactive atrial peptides (atriopeptins).
PMID 6419347
Abstract
Mammalian cardiac atria have several biologically active peptides that exert profound effects on sodium excretion, urine volume, and smooth muscle tone. In the present study two such peptides of low molecular weight were purified and separated from each other on the basis of differences in charge, hydrophobicity, and biological profile. The first peptide, designated atriopeptin I, exhibits natriuretic and diuretic activity and selectivity relaxes intestinal smooth muscle but not vascular smooth muscle strips. The second peptide, atriopeptin II, is a potent natriuretic and diuretic that relaxes both intestinal and vascular strips. Sequence analysis of atriopeptin I indicates that it is composed of 21 amino acids, of which serine and glycine residues predominate. The amino terminal sequence of atriopeptin II up to residue 21 is the same as that of atriopeptin I, with the addition of the Phe-Arg extension at the carboxyl terminus. Both peptides appear to be derived from a common high molecular weight precursor (designated atriopeptigen); their biological selectivity and potency may be determined by the site of carboxyl terminal cleavage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9035
Atriopeptin II rat
C98H156N34O32S2