EMAIL THIS PAGE TO A FRIEND
Clinical therapeutics
Aerosolized terbutaline sulfate in the treatment of bronchospasm.
PMID 6434183
Abstract
Terbutaline sulfate, a relatively selective beta2 agonist, is indicated for the treatment of bronchospasm associated with chronic obstructive pulmonary disease. Studies have shown that when administered via an aerosol, terbutaline has a rapid onset, a prolonged duration of action, and a low incidence of systemic side effects. Because the drug is delivered directly to the bronchi, one can administer low doses of aerosolized terbutaline and achieve a bronchodilatory effect comparable to that achieved with higher doses of the oral form.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2528
Terbutaline hemisulfate salt
C12H19NO3 · 0.5H2O4S
T0050000
Terbutaline sulfate, European Pharmacopoeia (EP) Reference Standard
C12H19NO3 · 0.5H2O4S