EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
A lectin from elder (Sambucus nigra L.) bark.
PMID 6466312
Abstract
A lectin was isolated from elder (Sambucus nigra) bark by affinity chromatography on fetuin-agarose. It is a tetrameric molecule (Mr 140000) composed of two different subunits of Mr 34500 and 37500 respectively, held together by intramolecular disulphide bridges. The lectin is a glycoprotein and is especially rich in asparagine/aspartic acid, glutamine/glutamic acid, valine and leucine. It is also the first lectin isolated from a species belonging to the plant family Caprifoliaceae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L6890 Lectin from Sambucus nigra (elder), lyophilized powder