EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Effects of various 2-amino-6-alkyldithiopurines on brain specific [3H]diazepam binding.
PMID 6539599
Abstract
Various derivatives of 2-amino-6- methylthiopurine with substituents at the 6-position of purine were tested for their abilities to displace [3H]diazepam binding to rat brain membranes. The potency was dependent on the carbon chain-length in the 6-position of purine. Among the derivatives tested, 2-amino-6-n- pentyldithiopurine had the highest potency, with a Ki value of 0.92 microM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9546
2-Amino-6-methylmercaptopurine, ≥95%
C6H7N5S