EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Chemiluminescence enzyme immunoassay of 17 alpha-hydroxyprogesterone using glucose oxidase and bis(2,4,6-trichlorophenyl) oxalate - fluorescent dye system.
PMID 6754116