EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
D-Galactonate/D-fuconate dehydratase.
PMID 6759856
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
80837
D-Fuconic acid lithium salt, ≥98.5% (TLC)
C6H12O6 · xLi+