EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Synthesis of chromogenic substrates specific for human spleen fibrinolytic proteinase (SFP) and human leukocyte elastase (LE).
PMID 6762255