EMAIL THIS PAGE TO A FRIEND
Toxicology
Decreased utilization of [2-14C]orotic acid for the synthesis of cytidine nucleotides in rat liver after administration of alpha-hexachlorocyclohexane.
PMID 67662
Abstract
Administration of alpha-1,2,3,4,5,6-hexachlorocyclohexane (alpha-HCH) to rats decreased the utilization of [2-14C]orotic acid for the synthesis of liver cytidine nucleotides. The specific radioactivities of uridine components of the acid-soluble pool and rRNA increased during the first hours of treatment with the drug. Later on the specific radioactivities of uridine nucleotides remained unchanged, while those of cytidine components decreased gradually. Administration of hydrocortisone increased the incorporation of labelled orotic acid into rRNA cytidylic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
300683
α-1,2,3,4,5,6-Hexachlorocyclohexane, 99%
C6H6Cl6