EMAIL THIS PAGE TO A FRIEND
Biochemistry
Synthesis and characterization of halogenated derivatives of the ionophore A23187: enhanced calcium ion transport specificity by the 4-bromo derivative.
PMID 6797465