EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Ammonium 4-chloro-7-sulfobenzofurazan: a new fluorigenic thiol-specific reagent.
PMID 6805440
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1154
4-Chloro-7-sulfobenzofurazan ammonium salt
C6H6ClN3O4S